Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTNNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171142.1 | NP_001164613.1 | 11 - 12 | 1747 | 109 | |
XM_008254892.2 | XP_008253114.1 | 11 - 12 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |