Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171266.1 | NP_001164737.1 | 2 - 3 | 328 | 100 | |
XM_008269909.2 | XP_008268131.1 | 2 - 3 | 362 | 100 | ||
GenBank mRNA | D26153.1 | - | 2 - 3 | 328 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |