Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171285.1 | NP_001164756.1 | 2 - 3 | 224 | 80 | |
XM_008261385.2 | XP_008259607.1 | 4 - 5 | 649 | 80 | ||
XM_008261386.2 | XP_008259608.1 | 4 - 5 | 682 | 80 | ||
XM_017343649.1 | XP_017199138.1 | 4 - 5 | 350 | 80 | ||
XM_017343650.1 | XP_017199139.1 | 4 - 5 | 645 | 80 | ||
XM_017343651.1 | XP_017199140.1 | 4 - 5 | 678 | 80 | ||
XM_017343652.1 | XP_017199141.1 | 4 - 5 | 355 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |