Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171341.1 | NP_001164812.1 | 4 - 5 | 1865 | 80 | |
XM_008262738.1 | XP_008260960.1 | 6 - 7 | 2154 | 80 | ||
XM_008262740.2 | XP_008260962.1 | 6 - 7 | 3016 | 80 | ||
XM_008262741.2 | XP_008260963.1 | 6 - 7 | 2496 | 80 | ||
XM_008262745.2 | XP_008260967.1 | 6 - 7 | 2026 | 80 | ||
XM_008262747.1 | XP_008260969.1 | 7 - 8 | 2072 | 80 | ||
XM_017344786.1 | XP_017200275.1 | 6 - 7 | 2125 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Oryctolagus cuniculus Probe spans exons |