Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171357.1 | NP_001164828.1 | 2 - 3 | 209 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |