Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171419.1 | 434 | 95 | |||
XM_008274925.2 | XP_008273147.1 | 633 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus |