Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195764.1 | NP_001182693.1 | 655 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus |