Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199820.1 | NP_001186749.1 | 335 | 92 | ||
GenBank mRNA | DN890943.1 | - | 608 | 92 | ||
EB379496.1 | - | 314 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus |