Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199841.1 | NP_001186770.1 | 9 - 10 | 1243 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus Probe spans exons |