Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199842.1 | NP_001186771.1 | 1 - 2 | 207 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Oryctolagus cuniculus Probe spans exons |