Search
Search

LOC4345375| Gene Symbol: LOC4345375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792679.2 | XP_015648165.1 | 1 - 2 | 759 | 105 | |
| GenBank mRNA | AK066365.1 | - | 2 - 3 | 704 | 105 | |
Set Membership: |
|