Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266571.2 | NP_001253500.1 | 44 | 63 | ||
XM_015146220.2 | XP_015001706.2 | 310 | 63 | |||
XM_015146222.2 | XP_015001708.2 | 70 | 63 | |||
XM_015146229.2 | XP_015001715.2 | 115 | 63 | |||
XM_015146230.2 | XP_015001716.2 | 427 | 63 | |||
XM_015146231.2 | XP_015001717.2 | 67 | 63 | |||
XM_015146232.2 | XP_015001718.2 | 69 | 63 | |||
XM_015146234.2 | XP_015001720.2 | 69 | 63 | |||
XM_015146236.2 | XP_015001722.2 | 197 | 63 | |||
XM_015146238.2 | XP_015001724.2 | 424 | 63 | |||
XM_015146239.2 | XP_015001725.2 | 143 | 63 | |||
XM_015146240.2 | XP_015001726.2 | 424 | 63 | |||
XM_028852269.1 | XP_028708102.1 | 109 | 63 | |||
GenBank mRNA | JU323957.1 | - | 110 | 63 | ||
JU472655.1 | - | 105 | 63 | |||
JV045382.1 | - | 44 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |