Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028841923.1 | XP_028697756.1 | 1779 | 78 | ||
XM_028841924.1 | XP_028697757.1 | 1618 | 78 | |||
GenBank mRNA | JV658507.1 | - | 484 | 78 | ||
JV720917.1 | - | 429 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |