Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPLP0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195428.1 | NP_001182357.1 | 277 | 76 | ||
XM_015152909.2 | XP_015008395.1 | 342 | 76 | |||
GenBank mRNA | CN642741.1 | - | 62 | 76 | ||
CN643487.1 | - | 277 | 76 | |||
JV047873.1 | - | 308 | 76 | |||
JV625067.1 | - | 322 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |