Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266768.1 | NP_001253697.1 | 1106 | 63 | ||
GenBank mRNA | JU332912.1 | - | 1031 | 63 | ||
JU332913.1 | - | 1031 | 63 | |||
JU474309.1 | - | 1106 | 63 | |||
JU474310.1 | - | 1106 | 63 | |||
JU474311.1 | - | 1106 | 63 | |||
JU474312.1 | - | 1106 | 63 | |||
JU474313.1 | - | 1106 | 63 | |||
JU474314.1 | - | 1106 | 63 | |||
JU474315.1 | - | 1106 | 63 | |||
JU474316.1 | - | 1106 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |