Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NONO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194913.2 | NP_001181842.1 | 1085 | 64 | ||
XM_001091198.4 | XP_001091198.1 | 1300 | 64 | |||
XM_001091316.4 | XP_001091316.1 | 1185 | 64 | |||
XM_028841688.1 | XP_028697521.1 | 663 | 64 | |||
GenBank mRNA | JU320152.1 | - | 804 | 64 | ||
JV637384.1 | - | 874 | 64 | |||
JV668356.1 | - | 787 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |