Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266139.1 | NP_001253068.1 | 728 | 130 | ||
GenBank mRNA | JV600209.1 | - | 728 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |