Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257790.2 | NP_001244719.1 | 2184 | 67 | ||
XM_015137117.2 | XP_014992603.1 | 2200 | 67 | |||
XM_015137119.2 | XP_014992605.1 | 2053 | 67 | |||
XM_015137121.2 | XP_014992607.1 | 2030 | 67 | |||
GenBank mRNA | JU333563.1 | - | 2185 | 67 | ||
JU474820.1 | - | 2112 | 67 | |||
JV046517.1 | - | 2043 | 67 | |||
JV710235.1 | - | 2189 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |