Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015128306.2 | XP_014983792.1 | 859 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |