Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257936.1 | NP_001244865.1 | 251 | 70 | ||
GenBank mRNA | JU320865.1 | - | 251 | 70 | ||
JU471786.1 | - | 176 | 70 | |||
JV047352.1 | - | 267 | 70 | |||
JV566392.1 | - | 254 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |