Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261807.1 | NP_001248736.1 | 471 | 83 | ||
XM_015114123.2 | XP_014969609.1 | 2502 | 83 | |||
XM_015114124.2 | XP_014969610.1 | 2591 | 83 | |||
GenBank mRNA | JU333382.1 | - | 541 | 83 | ||
JU474678.1 | - | 471 | 83 |