Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152082.2 | XP_015007568.1 | 592 | 125 | ||
GenBank mRNA | JU328574.1 | - | 477 | 125 | ||
JV635783.1 | - | 522 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |