Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118887.2 | XP_014974373.1 | 91 | |||
GenBank mRNA | AY680464.1 | - | 66 | 91 | ||
JU320404.1 | - | 91 | ||||
JV637368.1 | - | 91 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |