Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194274.2 | NP_001181203.1 | 616 | 75 | ||
XM_028853427.1 | XP_028709260.1 | 270 | 75 | |||
GenBank mRNA | JU320863.1 | - | 474 | 75 | ||
JV661984.1 | - | 414 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |