Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261131.1 | NP_001248060.1 | 537 | 67 | ||
GenBank mRNA | JU322329.1 | - | 564 | 67 | ||
JU470758.1 | - | 511 | 67 | |||
JU470759.1 | - | 511 | 67 | |||
JV048332.1 | - | 537 | 67 | |||
JV594826.1 | - | 573 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |