Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFE2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261099.2 | NP_001248028.1 | 1434 | 64 | ||
XM_015119374.2 | XP_014974860.1 | 1501 | 64 | |||
XM_015119377.2 | XP_014974863.1 | 793 | 64 | |||
GenBank mRNA | JU320594.1 | - | 1436 | 64 | ||
JV043404.1 | - | 1435 | 64 | |||
JV636580.1 | - | 1371 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |