Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261805.1 | NP_001248734.1 | 1383 | 53 | ||
XM_015144176.2 | XP_014999662.1 | 1635 | 53 | |||
GenBank mRNA | JU321019.1 | - | 1423 | 53 | ||
JV043956.1 | - | 1383 | 53 | |||
JV668465.1 | - | 1455 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |