Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257471.1 | NP_001244400.1 | 266 | 79 | ||
XM_015148713.2 | XP_015004199.1 | 416 | 79 | |||
GenBank mRNA | JU335346.1 | - | 266 | 79 | ||
JU476299.1 | - | 390 | 79 | |||
JU476300.1 | - | 390 | 79 | |||
JV043169.1 | - | 409 | 79 | |||
JV047580.1 | - | 409 | 79 | |||
JV637913.1 | - | 407 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |