Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258170.1 | NP_001245099.1 | 170 | 103 | ||
GenBank mRNA | JU321132.1 | - | 113 | 103 | ||
JU473799.1 | - | 170 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |