Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258046.1 | NP_001244975.1 | 998 | 62 | ||
XM_028832635.1 | XP_028688468.1 | 286 | 62 | |||
GenBank mRNA | JU474992.1 | - | 998 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |