Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266342.1 | NP_001253271.1 | 1231 | 131 | ||
GenBank mRNA | JU334559.1 | - | 1310 | 131 | ||
JU473764.1 | - | 1304 | 131 | |||
JV045164.1 | - | 1231 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |