Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152464.2 | XP_015007950.2 | 2199 | 142 | ||
XM_015152465.2 | XP_015007951.2 | 1921 | 142 | |||
XM_015152466.2 | XP_015007952.2 | 1716 | 142 | |||
XM_028829787.1 | XP_028685620.1 | 1577 | 142 | |||
GenBank mRNA | JU349354.1 | - | 1215 | 142 | ||
JU477461.1 | - | 1639 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |