Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836750.1 | XP_028692583.1 | 853 | 57 | ||
XM_028836752.1 | XP_028692585.1 | 839 | 57 | |||
XM_028836753.1 | XP_028692586.1 | 965 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |