Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265728.1 | NP_001252657.1 | 733 | 129 | ||
XM_028849187.1 | XP_028705020.1 | 1082 | 129 | |||
GenBank mRNA | JV565076.1 | - | 733 | 129 | ||
JV708763.1 | - | 710 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |