Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090833.4 | XP_001090833.3 | 995 | 57 | ||
XM_028826879.1 | XP_028682712.1 | 832 | 57 | |||
XM_028826880.1 | XP_028682713.1 | 720 | 57 | |||
XM_028826881.1 | XP_028682714.1 | 837 | 57 | |||
XM_028826882.1 | XP_028682715.1 | 694 | 57 | |||
GenBank mRNA | JV663075.1 | - | 745 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta Inventoried |