Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092797.4 | XP_001092797.3 | 1733 | 115 | ||
GenBank mRNA | JV047526.1 | - | 1756 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |