Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194119.3 | NP_001181048.1 | 1377 | 63 | ||
XM_001099323.4 | XP_001099323.1 | 1426 | 63 | |||
XM_015120818.2 | XP_014976304.1 | 1443 | 63 | |||
XM_015120819.2 | XP_014976305.1 | 1645 | 63 | |||
XM_015120820.2 | XP_014976306.1 | 1345 | 63 | |||
GenBank mRNA | JU336650.1 | - | 1329 | 63 | ||
JU473391.1 | - | 1355 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |