Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002806358.3 | XP_002806404.1 | 679 | 101 | ||
XM_015128025.2 | XP_014983511.1 | 679 | 101 | |||
XM_015128026.2 | XP_014983512.1 | 654 | 101 | |||
XM_028841834.1 | XP_028697667.1 | 679 | 101 | |||
XM_028841835.1 | XP_028697668.1 | 653 | 101 | |||
XM_028841836.1 | XP_028697669.1 | 679 | 101 | |||
XM_028841837.1 | XP_028697670.1 | 623 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |