Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137159.2 | XP_014992645.1 | 2241 | 156 | ||
GenBank mRNA | JU477459.1 | - | 2078 | 156 | ||
JV048415.1 | - | 2183 | 156 | |||
JV663152.1 | - | 608 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |