Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFR1OP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151148.2 | XP_015006634.1 | 860 | 68 | ||
GenBank mRNA | JU327933.1 | - | 33 | 68 | ||
JV045436.1 | - | 574 | 68 | |||
JV635518.1 | - | 786 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |