Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194773.2 | NP_001181702.1 | 394 | 81 | ||
XM_001092614.4 | XP_001092614.2 | 1082 | 81 | |||
XM_015111028.2 | XP_014966514.1 | 127 | 81 | |||
XM_028830499.1 | XP_028686332.1 | 106 | 81 | |||
XM_028830500.1 | XP_028686333.1 | 1086 | 81 | |||
GenBank mRNA | JU323188.1 | - | 199 | 81 | ||
JU475170.1 | - | 165 | 81 | |||
JV045482.1 | - | 227 | 81 | |||
JV636493.1 | - | 212 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |