Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257705.1 | NP_001244634.1 | 1271 | 64 | ||
XM_015143839.2 | XP_014999325.2 | 1494 | 64 | |||
XM_028850339.1 | XP_028706172.1 | 1347 | 64 | |||
XM_028850340.1 | XP_028706173.1 | 1046 | 64 | |||
GenBank mRNA | JU320672.1 | - | 1271 | 64 | ||
JU473133.1 | - | 1296 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |