Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194707.2 | NP_001181636.2 | 670 | 88 | ||
XM_028838537.1 | XP_028694370.1 | 705 | 88 | |||
GenBank mRNA | JU321878.1 | - | 664 | 88 | ||
JV044632.1 | - | 659 | 88 | |||
JV645032.1 | - | 674 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |