Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260909.1 | NP_001247838.1 | 544 | 73 | ||
XM_015125394.2 | XP_014980880.1 | 797 | 73 | |||
GenBank mRNA | JU320803.1 | - | 706 | 73 | ||
JU473074.1 | - | 544 | 73 | |||
JU473075.1 | - | 544 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |