Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015117133.2 | XP_014972619.1 | 1746 | 148 | ||
XM_015117134.2 | XP_014972620.1 | 1746 | 148 | |||
XM_015117135.2 | XP_014972621.1 | 1746 | 148 | |||
XM_015117136.2 | XP_014972622.1 | 1722 | 148 | |||
XM_015117137.2 | XP_014972623.1 | 1629 | 148 | |||
GenBank mRNA | JU326590.1 | - | 1636 | 148 | ||
JV647151.1 | - | 1680 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |