Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112692.2 | XP_014968178.1 | 684 | 96 | ||
XM_015112694.2 | XP_014968180.1 | 684 | 96 | |||
GenBank mRNA | JU325245.1 | - | 482 | 96 | ||
JU472254.1 | - | 1132 | 96 | |||
JV636321.1 | - | 2085 | 96 |