Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266697.1 | NP_001253626.1 | 511 | 67 | ||
XM_015135102.2 | XP_014990588.1 | 313 | 67 | |||
XM_015135106.2 | XP_014990592.1 | 574 | 67 | |||
XM_028844732.1 | XP_028700565.1 | 284 | 67 | |||
GenBank mRNA | JU333578.1 | - | 488 | 67 | ||
JV044516.1 | - | 306 | 67 | |||
JV528105.1 | - | 306 | 67 | |||
JV636051.1 | - | 511 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |