Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266697.1 | NP_001253626.1 | 2089 | 57 | ||
XM_015135102.2 | XP_014990588.1 | 1891 | 57 | |||
XM_015135106.2 | XP_014990592.1 | 2152 | 57 | |||
XM_028844732.1 | XP_028700565.1 | 1862 | 57 | |||
GenBank mRNA | JU333578.1 | - | 2066 | 57 | ||
JV044516.1 | - | 1884 | 57 | |||
JV528105.1 | - | 1884 | 57 | |||
JV636051.1 | - | 2089 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |