Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261760.1 | NP_001248689.1 | 991 | 98 | ||
XM_015141885.2 | XP_014997371.2 | 1811 | 98 | |||
XM_015141886.2 | XP_014997372.2 | 1806 | 98 | |||
XM_015141887.2 | XP_014997373.1 | 1226 | 98 | |||
XM_028850306.1 | XP_028706139.1 | 1095 | 98 | |||
GenBank mRNA | JU345278.1 | - | 923 | 98 | ||
JU476222.1 | - | 991 | 98 |