Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGIF2LX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032888.1 | NP_001028060.1 | 670 | 78 | ||
XM_015127766.2 | XP_014983252.2 | 648 | 78 | |||
GenBank mRNA | AJ345078.2 | - | 670 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |